RIE156322956 Rieke Metals, Inc.

2-chloro-4-fluorophenyl cyclobutyl ketone

  • MDL number MFCD03841279
  • Cas Number : 898791-03-2
  • Molecular Weight : 212.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-02-50

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-02-50
Availability details for
Quantity
  • Availability
  • Add to Cart