RIE156322960 Rieke Metals, Inc.

cyclobutyl 2,4-dichlorophenyl ketone

  • MDL number MFCD03841283
  • Cas Number : 898791-15-6
  • Molecular Weight : 229.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-02-54

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-02-54
Availability details for
Quantity
  • Availability
  • Add to Cart