RIE156322963 Rieke Metals, Inc.

Cyclobutyl 3,5-dichlorophenyl ketone

  • MDL number MFCD03841286
  • Cas Number : 898791-24-7
  • Molecular Weight : 229.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-02-57

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-02-57
Availability details for
Quantity
  • Availability
  • Add to Cart