RIE156322965 Rieke Metals, Inc.

Cyclobutyl 3,4-difluorophenyl ketone

  • MDL number MFCD03841288
  • Cas Number : 898791-28-1
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name cyclobutyl(3,4-difluorophenyl)methanone
Inchi Code 1S/C11H10F2O/c12-9-5-4-8(6-10(9)13)11(14)7-2-1-3-7/h4-7H,1-3H2
Inchi Key LMKGMUCWRQBMLX-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6123-02-59

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-02-59
Availability details for
Quantity
  • Availability
  • Add to Cart