RIE156322969 Rieke Metals, Inc.

cyclopentyl 3-methylphenyl ketone

  • MDL number MFCD03841292
  • Cas Number : 85359-50-8
  • Molecular Weight : 188.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-03-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-03-03
Availability details for
Quantity
  • Availability
  • Add to Cart