RIE156322973 Rieke Metals, Inc.

3-Cyanophenyl cyclopentyl ketone

  • MDL number MFCD03841298
  • Cas Number : 898791-36-1
  • Molecular Weight : 199.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-03-09

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-03-09
Availability details for
Quantity
  • Availability
  • Add to Cart