RIE156322977 Rieke Metals, Inc.

4-carboethoxyphenyl cyclopentyl ketone

  • MDL number MFCD03841302
  • Cas Number : 898791-40-7
  • Molecular Weight : 246.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-03-13

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-03-13
Availability details for
Quantity
  • Availability
  • Add to Cart