RIE156322979 Rieke Metals, Inc.

cyclopentyl 4-thiomethylphenyl ketone

  • MDL number MFCD03841304
  • Cas Number : 180048-75-3
  • Molecular Weight : 220.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-03-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-03-15
Availability details for
Quantity
  • Availability
  • Add to Cart