RIE156322982 Rieke Metals, Inc.

cyclopentyl 3-fluorophenyl ketone

  • MDL number MFCD03841309
  • Cas Number : 898791-46-3
  • Molecular Weight : 192.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-03-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-03-22
Availability details for
Quantity
  • Availability
  • Add to Cart