RIE156322983 Rieke Metals, Inc.

cyclopentyl 2,3-dimethylphenyl ketone

  • MDL number MFCD03841311
  • Cas Number : 898791-48-5
  • Molecular Weight : 202.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-03-26

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-03-26
Availability details for
Quantity
  • Availability
  • Add to Cart