RIE156322984 Rieke Metals, Inc.

cyclopentyl 2,4-dimethylphenyl ketone

  • MDL number MFCD03841312
  • Cas Number : 898791-50-9
  • Molecular Weight : 202.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-03-27

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-03-27
Availability details for
Quantity
  • Availability
  • Add to Cart