RIE156322985 Rieke Metals, Inc.

cyclopentyl 2,6-dimethylphenyl ketone

  • MDL number MFCD03841314
  • Cas Number : 898791-52-1
  • Molecular Weight : 202.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-03-29

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-03-29
Availability details for
Quantity
  • Availability
  • Add to Cart