RIE156322987 Rieke Metals, Inc.

4-bromo-3-fluorophenyl cyclopentyl ketone

  • MDL number MFCD03841317
  • Cas Number : 898791-58-7
  • Molecular Weight : 271.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-03-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-03-32
Availability details for
Quantity
  • Availability
  • Add to Cart