RIE156322989 Rieke Metals, Inc.

3-Chloro-4-fluorophenyl cyclopentyl ketone

  • MDL number MFCD03841319
  • Cas Number : 898791-62-3
  • Molecular Weight : 226.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-03-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-03-34
Availability details for
Quantity
  • Availability
  • Add to Cart