RIE156322992 Rieke Metals, Inc.

Cyclopentyl 3-trifluoromethylphenyl ketone

  • MDL number MFCD03841322
  • Cas Number : 898791-66-7
  • Molecular Weight : 242.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-03-44

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-03-44
Availability details for
Quantity
  • Availability
  • Add to Cart