RIE156322994 Rieke Metals, Inc.

4-bromo-2-fluorophenyl cyclopentyl ketone

  • MDL number MFCD03841324
  • Cas Number : 898791-68-9
  • Molecular Weight : 271.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-03-49

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-03-49
Availability details for
Quantity
  • Availability
  • Add to Cart