RIE156322996 Rieke Metals, Inc.

3-Chloro-5-fluorophenyl cyclopentyl ketone

  • MDL number MFCD03841326
  • Cas Number : 898791-72-5
  • Molecular Weight : 226.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-03-51

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-03-51
Availability details for
Quantity
  • Availability
  • Add to Cart