RIE156322998 Rieke Metals, Inc.

cyclopentyl 2,3-dichlorophenyl ketone

  • MDL number MFCD03841328
  • Cas Number : 898791-78-1
  • Molecular Weight : 243.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-03-53

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-03-53
Availability details for
Quantity
  • Availability
  • Add to Cart