RIE156323003 Rieke Metals, Inc.

cyclopentyl 3,4-difluorophenyl ketone

  • MDL number MFCD03841334
  • Cas Number : 898791-96-3
  • Molecular Weight : 210.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-03-59

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-03-59
Availability details for
Quantity
  • Availability
  • Add to Cart