RIE156323006 Rieke Metals, Inc.

Cyclohexyl 3-methoxyphenyl ketone

  • MDL number MFCD03841340
  • Cas Number : 898792-05-7
  • Molecular Weight : 218.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-04-06

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-04-06
Availability details for
Quantity
  • Availability
  • Add to Cart