RIE156323007 Rieke Metals, Inc.

2-cyanophenyl cyclohexyl ketone

  • MDL number MFCD03841342
  • Cas Number : 898792-08-0
  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-04-08

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-04-08
Availability details for
Quantity
  • Availability
  • Add to Cart