RIE156323009 Rieke Metals, Inc.

4-cyanophenyl cyclohexyl ketone

  • MDL number MFCD03841344
  • Cas Number : 898792-14-8
  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(cyclohexylcarbonyl)benzonitrile
Inchi Code 1S/C14H15NO/c15-10-11-6-8-13(9-7-11)14(16)12-4-2-1-3-5-12/h6-9,12H,1-5H2
Inchi Key PJPMXOBAIYBRRR-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6123-04-10

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-04-10
Availability details for
Quantity
  • Availability
  • Add to Cart