RIE156323010 Rieke Metals, Inc.

2-Carboethoxyphenyl cyclohexyl ketone

  • MDL number MFCD03841345
  • Cas Number : 898792-17-1
  • Molecular Weight : 260.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-04-11

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-04-11
Availability details for
Quantity
  • Availability
  • Add to Cart