RIE156323012 Rieke Metals, Inc.

4-carboethoxyphenyl cyclohexyl ketone

  • MDL number MFCD03841347
  • Cas Number : 684271-09-8
  • Molecular Weight : 260.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 4-(cyclohexylcarbonyl)benzoate
Inchi Code 1S/C16H20O3/c1-2-19-16(18)14-10-8-13(9-11-14)15(17)12-6-4-3-5-7-12/h8-12H,2-7H2,1H3
Inchi Key AEPOXFWOYIRIMT-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6123-04-13

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-04-13
Availability details for
Quantity
  • Availability
  • Add to Cart