RIE156323019 Rieke Metals, Inc.

cyclohexyl 3,5-dimethylphenyl ketone

  • MDL number MFCD03841359
  • Cas Number : 898769-18-1
  • Molecular Weight : 216.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-04-31

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-04-31
Availability details for
Quantity
  • Availability
  • Add to Cart