RIE156323022 Rieke Metals, Inc.

3-Chloro-4-fluorophenyl cyclohexyl ketone

  • MDL number MFCD03841362
  • Cas Number : 898769-27-2
  • Molecular Weight : 240.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-04-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-04-34
Availability details for
Quantity
  • Availability
  • Add to Cart