RIE156323026 Rieke Metals, Inc.

cyclohexyl 4-trifluoromethylphenyl ketone

  • MDL number MFCD03841367
  • Cas Number : 419543-02-5
  • Molecular Weight : 256.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name cyclohexyl[4-(trifluoromethyl)phenyl]methanone
Inchi Code 1S/C14H15F3O/c15-14(16,17)12-8-6-11(7-9-12)13(18)10-4-2-1-3-5-10/h6-10H,1-5H2
Inchi Key DPCFQJYEUCIZHF-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6123-04-45

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-04-45
Availability details for
Quantity
  • Availability
  • Add to Cart