RIE156323027 Rieke Metals, Inc.

4-bromo-2-fluorophenyl cyclohexyl ketone

  • MDL number MFCD03841368
  • Cas Number : 684271-18-9
  • Molecular Weight : 285.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-bromo-2-fluorophenyl)(cyclohexyl)methanone
Inchi Code 1S/C13H14BrFO/c14-10-6-7-11(12(15)8-10)13(16)9-4-2-1-3-5-9/h6-9H,1-5H2
Inchi Key QSWUMAHHKQNSGY-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6123-04-49

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-04-49
Availability details for
Quantity
  • Availability
  • Add to Cart