RIE156323028 Rieke Metals, Inc.

2-Chloro-4-fluorophenyl cyclohexyl ketone

  • MDL number MFCD03841369
  • Cas Number : 898769-33-0
  • Molecular Weight : 240.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-04-50

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-04-50
Availability details for
Quantity
  • Availability
  • Add to Cart