RIE156323030 Rieke Metals, Inc.

4-Chloro-2-fluorophenyl cyclohexyl ketone

  • MDL number MFCD03841371
  • Cas Number : 898769-39-6
  • Molecular Weight : 240.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-chloro-2-fluorophenyl)(cyclohexyl)methanone
Inchi Code 1S/C13H14ClFO/c14-10-6-7-11(12(15)8-10)13(16)9-4-2-1-3-5-9/h6-9H,1-5H2
Inchi Key LICGCLBTRKIORN-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6123-04-52

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-04-52
Availability details for
Quantity
  • Availability
  • Add to Cart