RIE156323034 Rieke Metals, Inc.

Cyclohexyl 3,5-dichlorophenyl ketone

  • MDL number MFCD03841376
  • Cas Number : 898769-51-2
  • Molecular Weight : 257.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-04-57

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-04-57
Availability details for
Quantity
  • Availability
  • Add to Cart