RIE156323035 Rieke Metals, Inc.

cyclohexyl 2,4-difluorophenyl ketone

  • MDL number MFCD03841377
  • Cas Number : 898769-54-5
  • Molecular Weight : 224.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-04-58

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-04-58
Availability details for
Quantity
  • Availability
  • Add to Cart