RIE156323039 Rieke Metals, Inc.

4-(morpholinomethyl)benzophenone

  • MDL number MFCD03841381
  • Cas Number : 789427-08-3
  • Molecular Weight : 281.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0100-01

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0100-01
Availability details for
Quantity
  • Availability
  • Add to Cart