RIE156323051 Rieke Metals, Inc.

4-carboethoxy-4'-morpholinomethyl benzophenone

  • MDL number MFCD03841393
  • Cas Number : 898769-84-1
  • Molecular Weight : 353.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0100-13

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0100-13
Availability details for
Quantity
  • Availability
  • Add to Cart