RIE156323053 Rieke Metals, Inc.

4-morpholinomethyl-4'-thiomethylbenzophenone

  • MDL number MFCD03841395
  • Cas Number : 898769-88-5
  • Molecular Weight : 327.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0100-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0100-15
Availability details for
Quantity
  • Availability
  • Add to Cart