RIE156323056 Rieke Metals, Inc.

3-chloro-4'-morpholinomethyl benzophenone

  • MDL number MFCD03841398
  • Cas Number : 898769-94-3
  • Molecular Weight : 315.80

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0100-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0100-20
Availability details for
Quantity
  • Availability
  • Add to Cart