RIE156323057 Rieke Metals, Inc.

4-chloro-4'-morpholinomethyl benzophenone

  • MDL number MFCD03841399
  • Cas Number : 898769-96-5
  • Molecular Weight : 315.80

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0100-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0100-21
Availability details for
Quantity
  • Availability
  • Add to Cart