RIE156323059 Rieke Metals, Inc.

4-fluoro-4'-morpholinomethyl benzophenone

  • MDL number MFCD03841401
  • Cas Number : 898770-00-8
  • Molecular Weight : 299.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-fluorophenyl)[4-(4-morpholinylmethyl)phenyl]methanone
Inchi Code 1S/C18H18FNO2/c19-17-7-5-16(6-8-17)18(21)15-3-1-14(2-4-15)13-20-9-11-22-12-10-20/h1-8H,9-13H2
Inchi Key MSMWDSCONADHTH-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0100-23

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0100-23
Availability details for
Quantity
  • Availability
  • Add to Cart