RIE156323064 Rieke Metals, Inc.

3,4-dimethyl-4'-morpholinomethyl benzophenone

  • MDL number MFCD03841406
  • Cas Number : 898770-14-4
  • Molecular Weight : 309.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0100-30

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0100-30
Availability details for
Quantity
  • Availability
  • Add to Cart