RIE156323065 Rieke Metals, Inc.

3,5-dimethyl-4'-morpholinomethyl benzophenone

  • MDL number MFCD03841407
  • Cas Number : 898770-17-7
  • Molecular Weight : 309.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,5-dimethylphenyl)[4-(4-morpholinylmethyl)phenyl]methanone
Inchi Code 1S/C20H23NO2/c1-15-11-16(2)13-19(12-15)20(22)18-5-3-17(4-6-18)14-21-7-9-23-10-8-21/h3-6,11-13H,7-10,14H2,1-2H3
Inchi Key FBRMCWKZGFPWJG-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0100-31

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0100-31
Availability details for
Quantity
  • Availability
  • Add to Cart