RIE156323066 Rieke Metals, Inc.

4-bromo-3-fluoro-4'-morpholinomethyl benzophenone

  • MDL number MFCD03841408
  • Cas Number : 898770-20-2
  • Molecular Weight : 378.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0100-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0100-32
Availability details for
Quantity
  • Availability
  • Add to Cart