RIE156323072 Rieke Metals, Inc.

4'-morpholinomethyl-3-trifluoromethylbenzophenone

  • MDL number MFCD03841414
  • Cas Number : 898770-38-2
  • Molecular Weight : 349.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0100-44

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0100-44
Availability details for
Quantity
  • Availability
  • Add to Cart