RIE156323075 Rieke Metals, Inc.

2-chloro-4-fluoro-4'-morpholinomethyl benzophenone

  • MDL number MFCD03841417
  • Cas Number : 898770-47-3
  • Molecular Weight : 333.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0100-50

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0100-50
Availability details for
Quantity
  • Availability
  • Add to Cart