RIE156323077 Rieke Metals, Inc.

4-chloro-2-fluoro-4'-morpholinomethyl benzophenone

  • MDL number MFCD03841419
  • Cas Number : 898770-53-1
  • Molecular Weight : 333.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0100-52

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0100-52
Availability details for
Quantity
  • Availability
  • Add to Cart