RIE156323084 Rieke Metals, Inc.

t-Butyl 2-pyridyl ketone

  • MDL number MFCD06738273
  • Cas Number : 31595-32-1
  • Molecular Weight : 163.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5569-11

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5569-11
Availability details for
Quantity
  • Availability
  • Add to Cart