RIE156323087 Rieke Metals, Inc.

n-Butyl picolinate

  • MDL number MFCD00023479
  • Cas Number : 5340-88-5
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5569-02C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5569-02C
Availability details for
Quantity
  • Availability
  • Add to Cart