RIE156323088 Rieke Metals, Inc.

Hexyl picolinate

  • MDL number MFCD07699192
  • Cas Number : 5394-95-6
  • Molecular Weight : 207.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name hexyl 2-pyridinecarboxylate
Inchi Code 1S/C12H17NO2/c1-2-3-4-7-10-15-12(14)11-8-5-6-9-13-11/h5-6,8-9H,2-4,7,10H2,1H3
Inchi Key XNQFSCZLUXHABZ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5569-02e

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5569-02e
Availability details for
Quantity
  • Availability
  • Add to Cart