RIE156323089 Rieke Metals, Inc.

2-Cyclohexanoyloxazole

  • MDL number MFCD07699311
  • Cas Number : 898758-88-8
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5571-06E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5571-06E
Availability details for
Quantity
  • Availability
  • Add to Cart