RIE156323092 Rieke Metals, Inc.

6-Chloro-1-(4-fluorophenyl)-1-oxohexane

  • MDL number MFCD01319942
  • Cas Number : 61191-90-0
  • Molecular Weight : 228.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6016E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6016E
Availability details for
Quantity
  • Availability
  • Add to Cart