RIE156323093 Rieke Metals, Inc.

7-Chloro-1-(4-fluorophenyl)-1-oxoheptane

  • MDL number MFCD01319943
  • Cas Number : 17135-47-6
  • Molecular Weight : 242.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6016F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6016F
Availability details for
Quantity
  • Availability
  • Add to Cart